Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tpcn2
Entrez Gene ID
233979
Gene Name
two pore segment channel 2
Gene Aliases
BC025890, D830047E22Rik
Chromosome Location
Chr.7: 145181633 - 145283987 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146206.4 NP_666318.2 5 - 6 509 94
GenBank mRNA AK040854.1 - 5 - 6 537 94
AK052930.1 - 5 - 6 510 94
AK089490.1 - 5 - 6 504 94
AK170863.1 - 5 - 6 505 94
BC043472.1 - 1 - 2 87 94
BC141195.1 - 5 - 6 593 94

View Full Product Details