The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013797.5 | NP_038825.1 | 2 - 3 | 527 | 118 |
GenBank mRNA | AB031813.1 | - | 2 - 3 | 368 | 118 |
AF148218.1 | - | 2 - 3 | 351 | 118 | |
AF223067.1 | - | 2 - 3 | 361 | 118 | |
AK041125.1 | - | 2 - 3 | 375 | 118 | |
AK165360.1 | - | 2 - 3 | 382 | 118 | |
AY195868.1 | - | 2 - 3 | 360 | 118 | |
AY195869.1 | - | 2 - 3 | 360 | 118 | |
AY243579.1 | - | 2 - 3 | 360 | 118 | |
BC041147.1 | - | 2 - 3 | 310 | 118 |