Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt33a
Entrez Gene ID
71888
Gene Name
keratin 33A
Gene Aliases
2310015J09Rik
Chromosome Location
Chr.11: 100011199 - 100016212 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027983.3 NP_082259.2 6 - 7 1160 113
GenBank mRNA AK009363.1 - 6 - 7 1161 113
AK037251.1 - 6 - 7 1161 113
BC029257.1 - 6 - 7 1143 113

View Full Product Details