The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290569.1 | NP_001277498.1 | 7 - 8 | 879 | 97 |
NM_029377.2 | NP_083653.1 | 7 - 8 | 753 | 97 | |
GenBank mRNA | AK007858.1 | - | 7 - 8 | 754 | 97 |
AY428767.1 | - | 7 - 8 | 603 | 97 | |
BC013546.1 | - | 7 - 8 | 735 | 97 |