The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134176.1 | NP_598937.1 | 1 - 1 | 277 | 128 |
GenBank mRNA | AK082576.1 | - | 5 - 5 | 927 | 128 |
BC132386.1 | - | 1 - 1 | 324 | 128 | |
BC138489.1 | - | 1 - 1 | 324 | 128 |