The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025957.4 | NP_080233.1 | 2 - 3 | 506 | 107 |
GenBank mRNA | AK005390.1 | - | 2 - 3 | 507 | 107 |
AK005482.1 | - | 2 - 3 | 507 | 107 | |
AK005533.1 | - | 2 - 3 | 507 | 107 | |
BC099412.1 | - | 2 - 3 | 436 | 107 |