The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008184.3 | NP_032210.3 | 1 - 2 | 150 | 134 |
GenBank mRNA | AJ000413.1 | - | 1 - 2 | 34 | 134 |
AK015265.1 | - | 1 - 2 | 136 | 134 | |
BC031818.1 | - | 1 - 2 | 113 | 134 | |
CB573365.1 | - | 1 - 2 | 153 | 134 |