The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013587.2 | NP_038615.2 | 6 - 7 | 847 | 85 |
GenBank mRNA | AJ639614.1 | - | 6 - 7 | 854 | 85 |
AK089250.1 | - | 6 - 7 | 850 | 85 | |
AK145710.1 | - | 6 - 7 | 890 | 85 | |
AK150451.1 | - | 6 - 7 | 860 | 84 | |
AK152485.1 | - | 6 - 7 | 860 | 84 | |
AK153323.1 | - | 6 - 7 | 860 | 84 | |
AK166616.1 | - | 6 - 7 | 850 | 85 | |
AK166746.1 | - | 6 - 7 | 861 | 85 | |
AK166819.1 | - | 6 - 7 | 865 | 85 | |
AK168421.1 | - | 6 - 7 | 850 | 85 | |
AK169132.1 | - | 6 - 7 | 855 | 85 | |
AK171471.1 | - | 6 - 7 | 850 | 85 | |
AK193187.1 | - | 3 - 4 | 275 | 85 | |
BC032170.1 | - | 6 - 7 | 850 | 85 | |
BC046641.1 | - | 6 - 7 | 839 | 85 | |
BC057979.1 | - | 6 - 7 | 818 | 85 | |
BC059887.1 | - | 6 - 7 | 664 | 85 | |
BC094324.1 | - | 6 - 7 | 847 | 85 | |
D00622.1 | - | 6 - 7 | 840 | 85 | |
S67967.1 | - | 6 - 7 | 873 | 85 |