Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rsl1d1
Entrez Gene ID
66409
Gene Name
ribosomal L1 domain containing 1
Gene Aliases
2410005K20Rik, C76439, pBK1
Chromosome Location
Chr.16: 11193037 - 11203292 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025546.2 NP_079822.1 5 - 6 664 71
GenBank mRNA AK012937.1 - 5 - 6 657 71
AK075933.1 - 5 - 6 665 71
AK136025.1 - 5 - 6 631 71
AK145699.1 - 5 - 6 678 71
AK162241.1 - 5 - 6 638 71
AK165572.1 - 5 - 6 71
AK166579.1 - 5 - 6 660 71
AK168469.1 - 5 - 6 660 71
AK168717.1 - 5 - 6 653 71
AK171805.1 - 5 - 6 652 71
BC034355.1 - 5 - 6 630 71
BC069934.1 - 5 - 6 638 71
DQ445383.1 - 5 - 6 672 71

View Full Product Details