The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026277.3 | NP_080553.1 | 4 - 5 | 409 | 62 |
GenBank mRNA | AK021194.1 | - | 4 - 5 | 413 | 62 |
AK075755.1 | - | 4 - 5 | 410 | 62 | |
BC034900.1 | - | 4 - 5 | 410 | 62 | |
BC041150.1 | - | 2 - 3 | 170 | 62 | |
BC054835.1 | - | 4 - 5 | 363 | 62 | |
BC103793.1 | - | 4 - 5 | 412 | 62 | |
BC117992.2 | - | 4 - 5 | 410 | 62 | |
BC117993.2 | - | 4 - 5 | 410 | 62 |