The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028295.1 | NP_082571.1 | 16 - 17 | 1578 | 68 |
GenBank mRNA | AK012415.1 | - | 4 - 5 | 369 | 68 |
BC009151.1 | - | 16 - 17 | 1578 | 68 |