Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Morc4
Entrez Gene ID
75746
Gene Name
microrchidia 4
Gene Aliases
1600017G11Rik, 5630401M14Rik, AA117449, Zcwcc2
Chromosome Location
Chr.X: 139821635 - 139871692 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193309.1 NP_001180238.1 15 - 16 2635 107
NM_029413.4 NP_083689.2 15 - 16 2635 107
GenBank mRNA AK032807.1 - 15 - 16 2636 107
AK160137.1 - 1 - 2 74 107
BC036164.1 - 9 - 10 1521 107
BC039283.1 - 8 - 9 1347 107
BC098483.1 - 15 - 16 2590 107
BC132153.1 - 15 - 16 2489 107
BC132497.1 - 15 - 16 2489 107

View Full Product Details