Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phf12
Entrez Gene ID
268448
Gene Name
PHD finger protein 12
Gene Aliases
2410142K10Rik, F630045O13, PF1, mKIAA1523
Chromosome Location
Chr.11: 77982754 - 78030539 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174852.3 NP_777277.2 2 - 3 742 59
GenBank mRNA AK010807.1 - 2 - 3 801 59
AK053787.1 - 2 - 3 743 59
AK089226.1 - 2 - 3 806 59
AK141796.1 - 2 - 3 646 59
AK155354.1 - 2 - 3 783 59
AK170155.1 - 2 - 3 830 59
AK170187.1 - 1 - 2 37 59
BC043080.1 - 2 - 3 345 59

View Full Product Details