Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Agap2
Entrez Gene ID
216439
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 2
Gene Aliases
AGAP-2, Centg1, PIKE, cnt-g1, mKIAA0167
Chromosome Location
Chr.10: 127075258 - 127093173 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033263.5 NP_001028435.1 2 - 3 1923 72
NM_001301014.1 NP_001287943.1 2 - 3 1923 72
GenBank mRNA AK147446.1 - 2 - 3 1924 72
AK220325.1 - 2 - 3 798 72

View Full Product Details