The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021510.2 | NP_067485.1 | 11 - 12 | 1334 | 68 |
GenBank mRNA | AK052983.1 | - | 9 - 10 | 3721 | 68 |
AK088169.1 | - | 11 - 12 | 1287 | 68 | |
AK161491.1 | - | - | 1432 | 68 | |
AK165283.1 | - | 6 - 7 | 679 | 68 | |
BC016459.1 | - | 9 - 10 | 3735 | 68 | |
BC027002.1 | - | 9 - 10 | 2186 | 68 | |
BC042187.1 | - | 11 - 12 | 1269 | 68 | |
BC056224.1 | - | 10 - 11 | 1264 | 68 | |
CO428102.1 | - | 3 - 4 | 170 | 68 |