The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029879.2 | NP_084155.2 | 2 - 3 | 1053 | 71 |
GenBank mRNA | AK020910.1 | - | 2 - 3 | 650 | 71 |
AK028323.1 | - | 2 - 3 | 430 | 71 | |
AK032138.1 | - | 2 - 3 | 1053 | 71 | |
AK036240.1 | - | 2 - 3 | 960 | 71 | |
AK046733.1 | - | 2 - 3 | 682 | 71 | |
AK137417.1 | - | 2 - 3 | 927 | 71 | |
BC118957.2 | - | 2 - 3 | 585 | 71 | |
DQ104214.1 | - | 2 - 3 | 343 | 71 |