Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cela1
Entrez Gene ID
109901
Gene Name
chymotrypsin-like elastase family, member 1
Gene Aliases
1810009A17Rik, 1810062B19Rik, Ela-1, Ela1, PC-TsF
Chromosome Location
Chr.15: 100674422 - 100687920 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033612.2 NP_291090.2 4 - 5 373 97
GenBank mRNA AK007392.1 - 4 - 5 371 97
BC011218.1 - 4 - 5 347 97
CB951068.1 - 4 - 5 373 97
M27347.1 - 3 - 4 256 97

View Full Product Details