Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sspo
Entrez Gene ID
243369
Gene Name
SCO-spondin
Gene Aliases
C79529, Scospondin
Chromosome Location
Chr.6: 48446243 - 48501262 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173428.3 NP_775604.3 60 - 61 9191 95
GenBank mRNA AJ491857.1 - 57 - 58 8756 95

View Full Product Details