The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177185.4 | NP_796159.3 | 14 - 15 | 3819 | 76 |
GenBank mRNA | AK051609.1 | - | 14 - 15 | 3192 | 76 |
AK173339.1 | - | 2 - 3 | 4855 | 76 | |
BC051458.1 | - | 14 - 15 | 3791 | 76 |