The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177185.4 | NP_796159.3 | 15 - 16 | 4048 | 60 |
GenBank mRNA | AK051609.1 | - | 15 - 16 | 3421 | 60 |
AK173339.1 | - | 3 - 4 | 5084 | 60 | |
BC051458.1 | - | 15 - 16 | 4020 | 60 |