Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Commd10
Entrez Gene ID
69456
Gene Name
COMM domain containing 10
Gene Aliases
2310003A05Rik, AU019438, DRWMS2
Chromosome Location
Chr.18: 46958785 - 47087995 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178377.4 NP_848464.1 3 - 4 250 67
GenBank mRNA AB052770.1 - 3 - 4 263 67
AK139321.1 - - 266 67
AK159099.1 - 3 - 4 266 67
AK166907.1 - 3 - 4 264 67
BC034879.1 - 3 - 4 250 67

View Full Product Details