Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Commd10
Entrez Gene ID
69456
Gene Name
COMM domain containing 10
Gene Aliases
2310003A05Rik, AU019438, DRWMS2
Chromosome Location
Chr.18: 46958785 - 47087995 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178377.4 NP_848464.1 4 - 5 409 73
GenBank mRNA AB052770.1 - 4 - 5 422 73
AK139321.1 - - 425 73
AK159099.1 - 4 - 5 425 73
AK166907.1 - 4 - 5 423 73
BC034879.1 - 4 - 5 409 73

View Full Product Details