Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agbl2
Entrez Gene ID
271813
Gene Name
ATP/GTP binding protein-like 2
Gene Aliases
4930524K04, A430081C19Rik, CCP2
Chromosome Location
Chr.2: 90782587 - 90834437 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178755.3 NP_848870.2 11 - 12 1963 67
GenBank mRNA AK029744.1 - 11 - 12 1996 67
AK029932.1 - 11 - 12 1964 67
AK030138.1 - 11 - 12 1951 67
AK030182.1 - 11 - 12 2010 67
AK040259.1 - 11 - 12 1899 67
AK041286.1 - 3 - 4 372 67
AK079691.1 - 11 - 12 1900 67
DQ867026.1 - 11 - 12 1996 67
DQ867027.1 - 11 - 12 1996 67
DQ867028.1 - 11 - 12 1996 67
DQ867029.1 - 11 - 12 1996 67
DQ867030.1 - 10 - 11 1942 67

View Full Product Details