Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tm6sf2
Entrez Gene ID
107770
Gene Name
transmembrane 6 superfamily member 2
Gene Aliases
-
Chromosome Location
Chr.8: 70072924 - 70080066 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293795.1 NP_001280724.1 7 - 8 782 99
NM_181540.4 NP_853518.1 7 - 8 776 99
GenBank mRNA AK077126.1 - 7 - 8 769 99
BC024498.1 - 7 - 8 770 99

View Full Product Details