The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293795.1 | NP_001280724.1 | 9 - 10 | 997 | 62 |
NM_181540.4 | NP_853518.1 | 9 - 10 | 991 | 62 | |
GenBank mRNA | AK077126.1 | - | 9 - 10 | 984 | 62 |
BC024498.1 | - | 9 - 10 | 985 | 62 |