Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sirt6
Entrez Gene ID
50721
Gene Name
sirtuin 6
Gene Aliases
2810449N18Rik, AI043036, Sir2l6
Chromosome Location
Chr.10: 81621786 - 81627792 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163430.1 NP_001156902.1 2 - 3 555 77
NM_181586.3 NP_853617.1 2 - 3 261 77
GenBank mRNA AK013316.1 - 2 - 3 210 77
AK145929.1 - 2 - 3 555 77
BC052763.1 - 2 - 3 211 77

View Full Product Details