The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181593.2 | NP_853624.1 | 2 - 3 | 1351 | 68 |
GenBank mRNA | AK033605.1 | - | 2 - 3 | 1020 | 68 |
AK154888.1 | - | 2 - 3 | 1383 | 68 | |
BC013227.1 | - | 2 - 3 | 1237 | 68 | |
BC053450.1 | - | 2 - 3 | 1351 | 68 |