Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Muc6
Entrez Gene ID
353328
Gene Name
mucin 6, gastric
Gene Aliases
-
Chromosome Location
Chr.7: 141634049 - 141655308 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181729.2 NP_859418.1 - 1023 80
GenBank mRNA BC120907.2 - 7 - 8 911 80

View Full Product Details