Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Krt85
Entrez Gene ID
53622
Gene Name
keratin 85
Gene Aliases
AA589597, BB007925, Hb5, Krt2-18
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016879.2 NP_058575.2 6 - 6 1050 86
GenBank mRNA AF021836.1 - 6 - 6 1034 86
AK009099.1 - 6 - 6 1049 86
AK028825.1 - 6 - 6 1050 86

View Full Product Details