The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147041.2 | NP_667252.1 | 1 - 1 | 981 | 94 |
GenBank mRNA | BC141307.1 | - | 1 - 1 | 985 | 94 |