The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277121.1 | NP_001264050.1 | 7 - 8 | 969 | 148 |
NM_001277122.1 | NP_001264051.1 | 6 - 7 | 808 | 148 | |
NM_001277123.1 | NP_001264052.1 | 6 - 7 | 803 | 148 | |
NM_028871.2 | NP_083147.1 | 7 - 8 | 972 | 148 | |
GenBank mRNA | AF441128.1 | - | 6 - 7 | 812 | 148 |
AK011307.1 | - | 6 - 7 | 721 | 148 | |
AK133840.1 | - | 6 - 7 | 729 | 148 | |
AK144992.1 | - | 6 - 7 | 726 | 148 | |
AK145849.1 | - | 6 - 7 | 747 | 148 | |
AK147043.1 | - | 7 - 8 | 938 | 148 | |
AK152039.1 | - | 7 - 8 | 944 | 148 | |
BC004679.1 | - | 6 - 7 | 720 | 148 | |
BC026850.1 | - | 7 - 8 | 903 | 148 | |
BC038051.1 | - | 7 - 8 | 949 | 148 |