Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srxn1
Entrez Gene ID
76650
Gene Name
sulfiredoxin 1 homolog (S. cerevisiae)
Gene Aliases
1700127B04Rik, AI854065, AW488194, Npn3, Srx, Srx1, TX01
Chromosome Location
Chr.2: 152105524 - 152111376 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029688.5 NP_083964.2 1 - 2 425 87
GenBank mRNA AK007296.1 - 1 - 2 220 87
AK089976.1 - 1 - 2 222 87
AK159226.1 - 1 - 2 220 87
AK159613.1 - 1 - 2 219 87
AK161066.1 - 1 - 2 262 87
BC011325.1 - 1 - 2 239 87
BC049957.1 - 1 - 2 425 87

View Full Product Details