Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slco1b2
- Entrez Gene ID
-
28253
- Gene Name
- solute carrier organic anion transporter family, member 1b2
- Gene Aliases
-
7330442B20Rik, OATP-C, OATP2, Oatp1b2, Oatp4, Slc21a10, Slc21a6, lst-1, mlst-1
- Chromosome Location
-
Chr.6: 141629452 - 141686646 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_020495.1 |
NP_065241.1 |
10 - 11 |
1312 |
137 |
GenBank mRNA |
AB031959.1 |
- |
10 - 11 |
1312 |
137 |
View Full Product Details