The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164769.1 | NP_001158241.1 | 2 - 3 | 171 | 104 |
NM_001164772.1 | NP_001158244.1 | 2 - 3 | 171 | 104 | |
NM_013890.4 | NP_038918.3 | 2 - 3 | 171 | 104 | |
GenBank mRNA | AK004015.1 | - | 2 - 3 | 137 | 104 |
AK005103.1 | - | 2 - 3 | 136 | 104 | |
AK009893.1 | - | 1 - 2 | 193 | 104 | |
AK014858.1 | - | 2 - 3 | 156 | 104 | |
AK033129.1 | - | 2 - 3 | 125 | 104 | |
AK040764.1 | - | 2 - 3 | 138 | 104 | |
AK145405.1 | - | 2 - 3 | 138 | 104 | |
BC003834.1 | - | 2 - 3 | 257 | 104 | |
X54352.1 | - | 1 - 2 | 101 | 104 |