The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007502.4 | NP_031528.1 | 4 - 4 | 539 | 156 |
GenBank mRNA | AK032516.1 | - | 4 - 4 | 556 | 156 |
AK148495.1 | - | 2 - 2 | 244 | 156 | |
AK151700.1 | - | 4 - 4 | 556 | 156 | |
AK151841.1 | - | 4 - 4 | 555 | 156 | |
AK159532.1 | - | 4 - 4 | 556 | 156 | |
AK161282.1 | - | 4 - 4 | 571 | 156 | |
AK168273.1 | - | 4 - 4 | 555 | 156 | |
AK218119.1 | - | 2 - 2 | 136 | 156 | |
BC079916.1 | - | 4 - 4 | 544 | 156 | |
U59761.1 | - | 4 - 4 | 542 | 156 |