The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164745.1 | NP_001158217.1 | 6 - 6 | 2687 | 150 |
NM_008974.4 | NP_033000.1 | 6 - 6 | 2718 | 150 | |
GenBank mRNA | AK033092.1 | - | 5 - 5 | 2432 | 150 |
AK045093.1 | - | 5 - 5 | 2420 | 150 | |
AK135662.1 | - | 5 - 5 | 1925 | 150 | |
AK155895.1 | - | 7 - 7 | 2762 | 150 | |
AK160163.1 | - | 1 - 1 | 1310 | 150 | |
BC086794.1 | - | 5 - 5 | 2099 | 150 |