Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Col4a5
Entrez Gene ID
12830
Gene Name
collagen, type IV, alpha 5
Gene Aliases
-
Chromosome Location
Chr.X: 141475369 - 141689236 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163155.1 NP_001156627.1 27 - 28 2459 86
NM_007736.4 NP_031762.2 28 - 29 2287 86
GenBank mRNA AB041350.1 - 27 - 28 2317 86
AK133179.1 - 28 - 29 2288 86
BC016479.1 - 3 - 4 224 86
BC043317.1 - 3 - 4 227 86
BC082788.1 - 27 - 28 2298 86

View Full Product Details