The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008584.3 | NP_032610.1 | 2 - 3 | 974 | 131 |
GenBank mRNA | AK028352.1 | - | 2 - 3 | 975 | 131 |
AK031152.1 | - | 2 - 3 | 931 | 131 | |
AK142690.1 | - | 2 - 3 | 853 | 131 | |
BC002076.1 | - | 2 - 3 | 873 | 131 | |
Z16406.1 | - | 2 - 3 | 852 | 131 |