Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alb
Entrez Gene ID
11657
Gene Name
albumin
Gene Aliases
Alb-1, Alb1
Chromosome Location
Chr.5: 90460889 - 90476603 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009654.3 NP_033784.2 5 - 6 657 75
GenBank mRNA AJ011413.1 - 5 - 6 644 75
AJ457860.1 - 5 - 6 611 75
AK010025.1 - 5 - 6 646 75
AK050248.1 - 5 - 6 646 75
AK050644.1 - 4 - 5 515 75
AK131891.1 - 5 - 6 649 75
AK160487.1 - 5 - 6 649 75
BC024643.1 - 5 - 6 631 75
BC049971.1 - 5 - 6 631 75

View Full Product Details