Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cnga3
Entrez Gene ID
12790
Gene Name
cyclic nucleotide gated channel alpha 3
Gene Aliases
CNG3
Chromosome Location
Chr.1: 37218278 - 37263384 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282010.1 NP_001268939.1 2 - 3 274 70
NM_009918.2 NP_034048.1 1 - 2 333 70
GenBank mRNA AJ243933.1 - 1 - 2 342 70
BC035272.1 - 1 - 2 277 70
BC049145.1 - 1 - 2 276 70
CJ167467.1 - 2 - 3 216 70

View Full Product Details