The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145885.1 | NP_001139357.1 | 2 - 3 | 74 | |
NM_010029.2 | NP_034159.1 | 2 - 3 | 74 | ||
GenBank mRNA | AK014844.1 | - | 2 - 3 | 74 | |
AK133366.1 | - | 2 - 3 | 74 | ||
BC137601.1 | - | 1 - 2 | 92 | 74 | |
BC144760.1 | - | 1 - 2 | 92 | 74 | |
BE860399.1 | - | 2 - 3 | 74 | ||
JQ923486.1 | - | 1 - 2 | 66 | 74 |