The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010306.3 | NP_034436.1 | 1 - 2 | 264 | 104 |
GenBank mRNA | AK004566.1 | - | 1 - 2 | 258 | 104 |
AK077490.1 | - | 1 - 2 | 226 | 104 | |
AK133749.1 | - | 1 - 2 | 205 | 104 | |
AK167438.1 | - | 1 - 2 | 218 | 104 | |
AK168578.1 | - | 1 - 2 | 202 | 104 | |
AK169067.1 | - | 1 - 2 | 209 | 104 | |
AK169285.1 | - | 1 - 2 | 239 | 104 | |
BC003462.1 | - | 1 - 2 | 192 | 104 | |
BC020009.1 | - | 1 - 2 | 236 | 104 | |
BC041107.1 | - | 1 - 2 | 175 | 104 | |
BY126665.1 | - | 1 - 2 | 265 | 104 |