Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pstpip1
Entrez Gene ID
19200
Gene Name
proline-serine-threonine phosphatase-interacting protein 1
Gene Aliases
CD2BP1, def-2
Chromosome Location
Chr.9: 56078445 - 56128890 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011193.2 NP_035323.2 10 - 11 1176 57
GenBank mRNA AK037936.1 - 9 - 10 1978 57
AK088150.1 - 9 - 10 1978 57
BC096761.1 - 10 - 11 956 57
U87814.1 - 10 - 11 1016 57

View Full Product Details