The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011324.2 | NP_035454.2 | 11 - 12 | 2256 | 123 |
GenBank mRNA | AF112185.1 | - | 11 - 12 | 1743 | 123 |
AK140389.1 | - | 11 - 12 | 2257 | 123 | |
BC040814.1 | - | 11 - 12 | 1886 | 123 | |
BC049952.1 | - | 9 - 10 | 847 | 123 | |
BC133688.1 | - | 11 - 12 | 1737 | 123 | |
BC133689.1 | - | 11 - 12 | 1737 | 123 |