Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spint2
Entrez Gene ID
20733
Gene Name
serine protease inhibitor, Kunitz type 2
Gene Aliases
AL024025, C76321, HAI-2
Chromosome Location
Chr.7: 29256330 - 29282064 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011464.2 NP_035594.1 1 - 2 466 63
GenBank mRNA AF099016.1 - 1 - 2 399 63
AI227451.1 - 1 - 2 102 63

View Full Product Details