The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011578.3 | NP_035708.2 | 10 - 11 | 1998 | 73 |
GenBank mRNA | AF039601.1 | - | 10 - 11 | 1798 | 73 |
AK133234.1 | - | 8 - 9 | 1508 | 73 | |
BC070428.1 | - | 10 - 11 | 1923 | 73 |