Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cops6
Entrez Gene ID
26893
Gene Name
COP9 (constitutive photomorphogenic) homolog, subunit 6 (Arabidopsis thaliana)
Gene Aliases
Sgn3, VIP/MOV34
Chromosome Location
Chr.5: 138161102 - 138163984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012002.3 NP_036132.1 9 - 10 846 68
GenBank mRNA AF071315.1 - 9 - 10 883 68
AK146410.1 - 9 - 10 857 68
AK146775.1 - 9 - 10 859 68
BC004664.1 - 9 - 10 845 68
BC014286.1 - 9 - 10 850 68

View Full Product Details