The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013811.3 | NP_038839.2 | - | 2288 | 103 |
GenBank mRNA | AF356520.1 | - | - | 2288 | 103 |
AF356521.1 | - | - | 2288 | 103 | |
AF356522.1 | - | - | 2298 | 103 | |
AF356523.1 | - | - | 2298 | 103 | |
AF363577.1 | - | - | 2298 | 103 |