Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pigv
Entrez Gene ID
230801
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class V
Gene Aliases
B330013B03, D430024F16Rik
Chromosome Location
Chr.4: 133659914 - 133673997 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178698.5 NP_848813.3 2 - 3 456 60
GenBank mRNA AK046527.1 - 2 - 3 417 60
AK047322.1 - 2 - 3 254 60
AK052446.1 - 2 - 3 430 60
AK080858.1 - 2 - 3 410 60
BC055060.1 - 2 - 3 406 60

View Full Product Details