Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fez1
Entrez Gene ID
235180
Gene Name
fasciculation and elongation protein zeta 1 (zygin I)
Gene Aliases
UNC-76, UNC76
Chromosome Location
Chr.9: 36821944 - 36878926 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183171.4 NP_898994.2 5 - 6 984 72
GenBank mRNA BC029629.1 - 6 - 7 1089 72
DQ141674.1 - 5 - 6 988 72

View Full Product Details